siRNA Details
| Gene Symbol: Ccdc79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180958.3 | NP_851289.2 | 4 | 343 | ||
| GenBank mRNA | AB775896.1 | - | 2 | 124 | ||
| AK029754.1 | - | 4 | 344 | |||
| BC113771.1 | - | 3 | 667 | |||
| BC129814.1 | - | 3 | 668 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc79| Gene Symbol: Ccdc79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180958.3 | NP_851289.2 | 4 | 343 | ||
| GenBank mRNA | AB775896.1 | - | 2 | 124 | ||
| AK029754.1 | - | 4 | 344 | |||
| BC113771.1 | - | 3 | 667 | |||
| BC129814.1 | - | 3 | 668 | |||