siRNA Details
| Gene Symbol: Ccdc79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180958.3 | NP_851289.2 | 14 | 1811 | ||
| GenBank mRNA | AB775896.1 | - | 12 | 1589 | ||
| AK029754.1 | - | 14 | 1812 | |||
| AK030157.1 | - | 13 | 2125 | |||
| BC113771.1 | - | 11 | 1851 | |||
| BC129814.1 | - | 11 | 1855 | |||

Ccdc79
Genomic Map