siRNA Details
| Gene Symbol: Ccdc67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181816.2 | NP_861537.2 | 3 | 332 | ||
| GenBank mRNA | AK030153.1 | - | 3 | 315 | ||
| BC096547.1 | - | 2 | 225 | 30360184 | ||
| BY069891.1 | - | 3 | 333 | |||
| KC211186.1 | - | 2 | 176 | |||

Ccdc67
Genomic Map