siRNA Details
| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183029.2 | NP_898850.2 | 16 | 2463 | ||
| GenBank mRNA | AK044984.1 | - | 15 | 1979 | ||
| AK049196.1 | - | 16 | 2159 | |||
| AK159654.1 | - | 16 | 2196 | |||
| AK170531.1 | - | 15 | 1967 | |||
| BC023758.1 | - | 16 | 2161 | |||
| BC054552.1 | - | 15 | 3563 | |||

Igf2bp2
Genomic Map