siRNA Details
| Gene Symbol: Nup54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183392.2 | NP_899248.1 | 8, 9 | 1192 | ||
| GenBank mRNA | AK087827.1 | - | 1086 | |||
| AK088855.1 | - | 8, 9 | 1117 | |||
| AK133841.1 | - | 8, 9 | 1190 | |||
| AK150182.1 | - | 8, 9 | 1117 | |||
| AK150841.1 | - | 8, 9 | 1078 | |||
| AK151896.1 | - | 8, 9 | 1078 | |||
| AK152136.1 | - | 8, 9 | 1078 | |||
| AK162037.1 | - | 8, 9 | 1124 | |||
| BC044303.1 | - | 8, 9 | 1117 | |||
| BC068114.1 | - | 8, 9 | 1077 | 5707888 | ||

Nup54
Genomic Map