siRNA Details
| Gene Symbol: Sidt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159419.1 | NP_001152891.1 | 23 | 2453 | ||
| NM_198034.3 | NP_932151.2 | 22 | 2438 | |||
| GenBank mRNA | AK034344.1 | - | 8 | 603 | ||
| AK171920.1 | - | 22 | 2591 | |||
| AY562213.1 | - | 12 | 997 | |||
| AY562214.1 | - | 12 | 997 | |||
| BC025888.1 | - | 12 | 1088 | 5039343 | ||
| BC079584.1 | - | 22 | 2438 | 5704672 | ||

Sidt1
Genomic Map