siRNA Details
| Gene Symbol: Zbtb39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198035.1 | NP_932152.1 | 2 | 929 | ||
| GenBank mRNA | AK155700.1 | - | 2 | 905 | ||
| AK172936.1 | - | 1 | 682 | |||
| BC058661.1 | - | 2 | 929 | 6409441 | ||
| BC060725.1 | - | 1 | 538 | |||
| BC079569.1 | - | 2 | 825 | 30536951 | ||

Zbtb39
Genomic Map