siRNA Details
| Gene Symbol: Orai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198424.3 | NP_940816.1 | 2 | 1264 | ||
| GenBank mRNA | AK037138.1 | - | 3 | 1408 | ||
| AK136955.1 | - | 2 | 1265 | |||
| BC061259.1 | - | 2 | 1234 | 30251946 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orai3| Gene Symbol: Orai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198424.3 | NP_940816.1 | 2 | 1264 | ||
| GenBank mRNA | AK037138.1 | - | 3 | 1408 | ||
| AK136955.1 | - | 2 | 1265 | |||
| BC061259.1 | - | 2 | 1234 | 30251946 | ||