siRNA Details
| Gene Symbol: Tspyl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198617.2 | NP_941019.2 | 1 | 1192 | ||
| GenBank mRNA | AK134522.1 | - | 1 | 1189 | ||
| AK148882.1 | - | 1 | 1137 | |||
| AK167722.1 | - | 1 | 1173 | |||
| BC058340.1 | - | 1 | 1134 | 6590443 | ||

Tspyl3
Genomic Map