siRNA Details
| Gene Symbol: Xxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198626.2 | NP_941028.2 | 1 | 516 | ||
| GenBank mRNA | AK154256.1 | - | 1 | 516 | ||
| AK166259.1 | - | 1 | 469 | |||
| BC031419.1 | - | 1 | 464 | 2615706 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xxylt1| Gene Symbol: Xxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198626.2 | NP_941028.2 | 1 | 516 | ||
| GenBank mRNA | AK154256.1 | - | 1 | 516 | ||
| AK166259.1 | - | 1 | 469 | |||
| BC031419.1 | - | 1 | 464 | 2615706 | ||