siRNA Details
| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199021.3 | NP_950186.3 | 3 | 368 | ||
| GenBank mRNA | AK032579.1 | - | 1 | 14 | ||
| AK046842.1 | - | 3 | 370 | |||
| BC067026.1 | - | 3 | 361 | 6853947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpp10| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199021.3 | NP_950186.3 | 3 | 368 | ||
| GenBank mRNA | AK032579.1 | - | 1 | 14 | ||
| AK046842.1 | - | 3 | 370 | |||
| BC067026.1 | - | 3 | 361 | 6853947 | ||