siRNA Details
| Gene Symbol: Dpp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199021.3 | NP_950186.3 | 26 | 3865 | ||
| GenBank mRNA | AK032579.1 | - | 24 | 3511 | ||
| BC029696.1 | - | 11 | 2347 | |||
| BC063074.1 | - | 16 | 2787 | 5697531 | ||
| BC067026.1 | - | 26 | 3858 | 6853947 | ||

Dpp10
Genomic Map