siRNA Details
| Gene Symbol: Itgb1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008403.4 | NP_032429.1 | 7 | 754 | ||
| GenBank mRNA | AK002786.1 | - | 7 | 711 | ||
| AK004172.1 | - | 7 | 710 | |||
| AK007635.1 | - | 7 | 712 | |||
| AK075577.1 | - | 7 | 755 | |||
| AK089937.1 | - | 7 | 868 | |||
| AK210343.1 | - | 2 | 146 | |||
| BC028772.1 | - | 6 | 590 | 1230640 | ||

Itgb1bp1
Genomic Map