siRNA Details
| Gene Symbol: Kif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008445.2 | NP_032471.2 | 7 | 3018 | ||
| GenBank mRNA | AF013116.1 | - | 7 | 2594 | ||
| AK029381.1 | - | 1 | 418 | |||
| AK139894.1 | - | 7 | 694 | |||
| AK147572.1 | - | 7 | 2820 | |||
| AK220283.1 | - | 8 | 2743 | |||
| BC052732.1 | - | 7 | 2175 | |||
| BC053053.1 | - | 7 | 2960 | |||
| BC062648.1 | - | 7 | 2693 | |||
| BC127063.1 | - | 7 | 2230 | 40109965 | ||

Kif3c
Genomic Map