siRNA Details
| Gene Symbol: Klf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008452.2 | NP_032478.2 | 3 | 1425 | ||
| GenBank mRNA | AK131962.1 | - | 3 | 1426 | ||
| U25096.1 | - | 3 | 1452 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf2| Gene Symbol: Klf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008452.2 | NP_032478.2 | 3 | 1425 | ||
| GenBank mRNA | AK131962.1 | - | 3 | 1426 | ||
| U25096.1 | - | 3 | 1452 | |||