siRNA Details
| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008465.5 | NP_032491.2 | 13 | 1531 | ||
| GenBank mRNA | AK028259.1 | - | 13 | 1532 | ||
| AK028307.1 | - | 13 | 1550 | |||
| AK154800.1 | - | 13 | 1549 | |||
| AK169311.1 | - | 13 | 1554 | |||
| AK169348.1 | - | 13 | 1554 | |||
| AK203891.1 | - | 2 | 198 | |||
| BC006771.1 | - | 13 | 1516 | 3588534 | ||
| U20619.1 | - | 13 | 1536 | |||
| U34228.1 | - | 12 | 1380 | |||

Kpna1
Genomic Map