siRNA Details
| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008466.5 | NP_032492.1 | 3, 4 | 366 | ||
| GenBank mRNA | AF020772.1 | - | 3, 4 | 210 | ||
| AK011559.1 | - | 3, 4 | 237 | |||
| AK049373.1 | - | 3, 4 | 237 | |||
| AK161664.1 | - | 3, 4 | 294 | |||
| AV581906.1 | - | 3, 4 | 366 | |||
| BC026885.1 | - | 3, 4 | 260 | 3979788 | ||
| CJ063714.1 | - | 3, 4 | 451 | |||

Kpna3
Genomic Map