siRNA Details
| Gene Symbol: Ktn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293635.1 | NP_001280564.1 | 4 | 946 | ||
| NM_001293636.1 | NP_001280565.1 | 5 | 1066 | |||
| NM_008477.2 | NP_032503.2 | 3 | 817 | |||
| GenBank mRNA | AK040200.1 | - | 4 | 944 | ||
| BC066871.1 | - | 4 | 901 | |||
| BC075697.1 | - | 5 | 985 | |||
| CO431187.1 | - | 3 | 654 | |||
| L43326.1 | - | 3 | 805 | |||

Ktn1
Genomic Map