siRNA Details
| Gene Symbol: Lrmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281980.1 | NP_001268909.1 | 11 | 780 | ||
| NM_001281981.1 | NP_001268910.1 | 12 | 821 | |||
| NM_008511.3 | NP_032537.3 | 11 | 1052 | |||
| GenBank mRNA | AK040180.1 | - | 11 | 1053 | ||
| AK050646.1 | - | 11 | 1490 | |||
| AK079932.1 | - | 11 | 1010 | |||
| AK153823.1 | - | 11 | 1050 | |||
| AK153865.1 | - | 11 | 782 | |||
| AK160009.1 | - | 11 | 1012 | |||
| AK166635.1 | - | 9 | 501 | |||
| AK168775.1 | - | 9 | 478 | |||
| AK169578.1 | - | 12 | 805 | |||
| AY753324.1 | - | 10 | 615 | |||
| BC052909.1 | - | 11 | 988 | 30076553 | ||
| U10484.1 | - | 11 | 758 | |||

Lrmp
Genomic Map