siRNA Details
| Gene Symbol: Lrrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008516.4 | NP_032542.1 | 2 | 1020 | ||
| GenBank mRNA | AK139355.1 | - | 2 | 978 | ||
| AK139965.1 | - | 2 | 1022 | |||
| AK160841.1 | - | 2 | 899 | |||
| AK164849.1 | - | 2 | 979 | |||
| AK173186.1 | - | 1 | 265 | |||
| BC031122.1 | - | 1 | 295 | 4972803 | ||
| BC058701.1 | - | 2 | 953 | 6853964 | ||
| D45913.1 | - | 2 | 966 | |||

Lrrn1
Genomic Map