siRNA Details
| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008547.2 | NP_032573.2 | 11, 12 | 1816 | ||
| GenBank mRNA | X66983.1 | - | 11, 12 | 1810 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mak| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008547.2 | NP_032573.2 | 11, 12 | 1816 | ||
| GenBank mRNA | X66983.1 | - | 11, 12 | 1810 | ||