siRNA Details
| Gene Symbol: Mcpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008570.1 | NP_032596.1 | 4 | 398 | ||
| GenBank mRNA | BC100700.1 | - | 4 | 389 | 40043851 | |
| BC100701.1 | - | 4 | 389 | 40043850 | ||
| BC100702.1 | - | 4 | 389 | 40043849 | ||
| BC100703.1 | - | 4 | 389 | 40043848 | ||
| S44609.1 | - | 4 | 391 | |||

Mcpt1
Genomic Map