siRNA Details
| Gene Symbol: Mdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF007110.1 | - | 3, 4 | 326 | ||
| AK017719.1 | - | 3, 4 | 300 | |||
| AK049576.1 | - | 3, 4 | 291 | |||
| AK049690.1 | - | 3, 4 | 295 | |||
| AK083349.1 | - | 4, 5 | 361 | |||
| AK139529.1 | - | 3, 4 | 236 | |||
| AK139535.1 | - | 3, 4 | 234 | |||
| AK161882.1 | - | 273 | ||||
| BC003750.1 | - | 3, 4 | 305 | 3599906 | ||
| CJ101093.1 | - | 3, 4 | 350 | |||
| EU568360.1 | - | 4, 5 | 357 | |||

Mdm4
Genomic Map