siRNA Details
| Gene Symbol: Mdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF007110.1 | - | 4 | 375 | ||
| AK017719.1 | - | 4 | 349 | |||
| AK049576.1 | - | 4 | 340 | |||
| AK049690.1 | - | 4 | 344 | |||
| AK083349.1 | - | 5 | 410 | |||
| AK139529.1 | - | 4 | 285 | |||
| AK139535.1 | - | 4 | 283 | |||
| AK161882.1 | - | 322 | ||||
| AK184147.1 | - | 2 | 89 | |||
| BC003750.1 | - | 4 | 354 | 3599906 | ||
| CJ101093.1 | - | 4 | 399 | |||
| EU568360.1 | - | 5 | 406 | |||

Mdm4
Genomic Map