siRNA Details
| Gene Symbol: Mdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF007110.1 | - | 4 | 393 | ||
| AK017719.1 | - | 4 | 367 | |||
| AK049576.1 | - | 4 | 358 | |||
| AK049690.1 | - | 4 | 362 | |||
| AK083349.1 | - | 5 | 428 | |||
| AK139529.1 | - | 4 | 303 | |||
| AK139535.1 | - | 4 | 301 | |||
| AK161882.1 | - | 340 | ||||
| AK184147.1 | - | 2 | 107 | |||
| BC003750.1 | - | 4 | 372 | 3599906 | ||
| EU568360.1 | - | 5 | 424 | |||

Mdm4
Genomic Map