siRNA Details
| Gene Symbol: ITM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004867.4 | NP_004858.1 | 2 | 541 | ||
| GenBank mRNA | AB209310.1 | - | 1 | 3836 | ||
| AF038953.1 | - | 2 | 343 | |||
| AK223327.1 | - | 2 | 320 | |||
| AK313154.1 | - | 2 | 499 | |||
| AY359051.1 | - | 2 | 282 | |||
| BC040437.1 | - | 2 | 296 | 5748519 | ||
| CR456875.1 | - | 2 | 204 | |||

ITM2A
Genomic Map