siRNA Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008591.2 | NP_032617.2 | 3 | 1025 | ||
| GenBank mRNA | AK148258.1 | - | 3 | 876 | ||
| AK170560.1 | - | 3 | 1025 | |||
| BC117969.1 | - | 1 | 669 | |||
| BC117970.1 | - | 1 | 665 | |||
| Y00671.1 | - | 1 | 670 | |||

Met
Genomic Map