siRNA Details
| Gene Symbol: Mfge8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008594.2 | NP_032620.2 | 3, 4 | 427 | ||
| GenBank mRNA | AB021130.2 | - | 3, 4 | 334 | ||
| AK152088.1 | - | 3, 4 | 416 | |||
| AK155047.1 | - | 3, 4 | 425 | |||
| AK171143.1 | - | 3, 4 | 391 | |||
| AK195127.1 | - | 2, 3 | 269 | |||
| AK205061.1 | - | 2, 3 | 213 | |||
| M38337.1 | - | 3, 4 | 379 | |||

Mfge8
Genomic Map