siRNA Details
| Gene Symbol: Mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008595.2 | NP_032621.1 | 8 | 1570 | ||
| GenBank mRNA | AK143523.1 | - | 8 | 1604 | ||
| BC010983.1 | - | 8 | 1592 | 3980692 | ||
| U94349.1 | - | 8 | 1567 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfng| Gene Symbol: Mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008595.2 | NP_032621.1 | 8 | 1570 | ||
| GenBank mRNA | AK143523.1 | - | 8 | 1604 | ||
| BC010983.1 | - | 8 | 1592 | 3980692 | ||
| U94349.1 | - | 8 | 1567 | |||