siRNA Details
| Gene Symbol: Mitf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113198.1 | NP_001106669.1 | 5 | 829 | ||
| NM_001178049.1 | NP_001171520.1 | 5 | 716 | |||
| NM_008601.3 | NP_032627.1 | 4 | 520 | |||
| GenBank mRNA | AB221657.1 | - | 4 | 382 | ||
| AK052895.1 | - | 5 | 688 | |||
| AK154189.1 | - | 5 | 830 | |||
| AK155350.1 | - | 5 | 830 | |||
| AK200280.1 | - | 3 | 186 | |||
| BC108976.2 | - | 4 | 484 | 40047440 | ||
| BC108977.1 | - | 4 | 449 | 40047441 | ||
| Z23066.1 | - | 4 | 511 | |||

Mitf
Genomic Map