siRNA Details
| Gene Symbol: Mitf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113198.1 | NP_001106669.1 | 10 | 1909 | ||
| NM_001178049.1 | NP_001171520.1 | 10 | 1796 | |||
| NM_008601.3 | NP_032627.1 | 9 | 1600 | |||
| GenBank mRNA | AK154189.1 | - | 11 | 1911 | ||
| AK155350.1 | - | 10 | 1910 | |||
| BC108976.2 | - | 9 | 1564 | 40047440 | ||
| BC108977.1 | - | 9 | 1529 | 40047441 | ||
| Z23066.1 | - | 9 | 1591 | |||

Mitf
Genomic Map