siRNA Details
| Gene Symbol: Mmp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008609.3 | NP_032635.1 | 4 | 1226 | ||
| GenBank mRNA | BC047278.1 | - | 4 | 1226 | 6390113 | |
| BC057952.1 | - | 4 | 796 | 5342023 | ||
| D86332.1 | - | 4 | 722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp15| Gene Symbol: Mmp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008609.3 | NP_032635.1 | 4 | 1226 | ||
| GenBank mRNA | BC047278.1 | - | 4 | 1226 | 6390113 | |
| BC057952.1 | - | 4 | 796 | 5342023 | ||
| D86332.1 | - | 4 | 722 | |||