siRNA Details
| Gene Symbol: Mns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008613.3 | NP_032639.1 | 3 | 459 | ||
| GenBank mRNA | AK013048.1 | - | 3 | 426 | ||
| AK132772.1 | - | 3 | 434 | |||
| BC046624.1 | - | 3 | 372 | 6314904 | ||
| D14849.1 | - | 3 | 490 | |||

Mns1
Genomic Map