siRNA Details
| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039364.2 | NP_001034453.1 | 3 | 375 | ||
| NM_008614.2 | NP_032640.1 | 4 | 454 | |||
| GenBank mRNA | AK028057.1 | - | 4 | 454 | ||
| AK083103.1 | - | 4 | 455 | |||
| BY257495.1 | - | 3 | 376 | |||
| U81317.1 | - | 3 | 374 | |||

Mobp
Genomic Map