siRNA Details
| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039364.2 | NP_001034453.1 | 3 | 2514 | ||
| NM_008614.2 | NP_032640.1 | 4 | 2593 | |||
| GenBank mRNA | AK028057.1 | - | 4 | 2593 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mobp| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039364.2 | NP_001034453.1 | 3 | 2514 | ||
| NM_008614.2 | NP_032640.1 | 4 | 2593 | |||
| GenBank mRNA | AK028057.1 | - | 4 | 2593 | ||