siRNA Details
| Gene Symbol: Prol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008644.2 | NP_032670.2 | 3 | 851 | ||
| GenBank mRNA | AF247816.1 | - | 4 | 577 | ||
| BC021401.1 | - | 4 | 433 | 4166946 | ||
| BC042902.1 | - | 4 | 705 | 4924369 | ||
| U37531.1 | - | 4 | 734 | |||

Prol1
Genomic Map