siRNA Details
| Gene Symbol: Mybl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008652.2 | NP_032678.1 | 4 | 442 | ||
| GenBank mRNA | AK028497.1 | - | 4 | 442 | ||
| AK183670.1 | - | 1 | 35 | |||
| BC050842.1 | - | 4 | 388 | 6518298 | ||
| X70472.1 | - | 4 | 390 | |||

Mybl2
Genomic Map