siRNA Details
| Gene Symbol: MRPL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004891.3 | NP_004882.1 | 4 | 396 | ||
| NM_145330.2 | NP_663303.1 | 3 | 289 | |||
| GenBank mRNA | AF047440.1 | - | 4 | 370 | ||
| BC009475.1 | - | 4 | 348 | 4049053 | ||
| BC020834.1 | - | 4 | 383 | 4775045 | ||
| BC087846.1 | - | 3 | 253 | |||
| BC090047.1 | - | 3 | 289 | 30393466 | ||
| BI601959.1 | - | 4 | 399 | |||

MRPL33
Genomic Map