siRNA Details
| Gene Symbol: Gadd45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008655.1 | NP_032681.1 | 4 | 667 | ||
| GenBank mRNA | AK010420.1 | - | 4 | 685 | ||
| AK149895.1 | - | 4 | 685 | |||
| AK154952.1 | - | 4 | 685 | |||
| BC023815.1 | - | 4 | 679 | 5355150 | ||
| X54149.1 | - | 4 | 667 | |||

Gadd45b
Genomic Map