siRNA Details
| Gene Symbol: Nab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008667.3 | NP_032693.2 | 2, 3 | 1067 | ||
| GenBank mRNA | AB221677.1 | - | 1, 2 | 809 | ||
| BC005627.1 | - | 2, 3 | 1067 | 3155795 | ||
| BC016886.1 | - | 2, 3 | 990 | 4236565 | ||
| U47008.1 | - | 2, 3 | 945 | |||

Nab1
Genomic Map