siRNA Details
| Gene Symbol: Nab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122895.1 | NP_001116367.1 | 3 | 1248 | ||
| NM_008668.2 | NP_032694.2 | 3 | 1248 | |||
| GenBank mRNA | AK158688.1 | - | 3 | 1178 | ||
| AK171180.1 | - | 3 | 1128 | |||
| BC045139.1 | - | 3 | 1132 | 6306757 | ||
| U47543.1 | - | 3 | 1223 | |||

Nab2
Genomic Map