siRNA Details
| Gene Symbol: Nap1l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285489.1 | NP_001272418.1 | 9 | 803 | ||
| NM_001285490.1 | NP_001272419.1 | 9 | 816 | |||
| NM_008672.3 | NP_032698.1 | 9 | 803 | |||
| GenBank mRNA | AJ002198.1 | - | 9 | 773 | ||
| AK090112.1 | - | 9 | 819 | |||
| AK156835.1 | - | 9 | 776 | |||
| BC089018.1 | - | 9 | 789 | |||
| BC132039.1 | - | 8 | 727 | 40130316 | ||
| BC138518.1 | - | 8 | 727 | 8861538 | ||
| BC145306.1 | - | 8 | 727 | 9053860 | ||
| BC145308.1 | - | 8 | 727 | 9053862 | ||

Nap1l4
Genomic Map