siRNA Details
| Gene Symbol: Nefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008691.2 | NP_032717.2 | 1 | 957 | ||
| GenBank mRNA | AK051296.1 | - | 1 | 800 | ||
| AK051696.1 | - | 1 | 797 | |||
| AK080794.1 | - | 1 | 801 | |||
| AK164318.1 | - | 1 | 800 | |||
| AK165041.1 | - | 1 | 800 | |||
| BC052352.1 | - | 1 | 799 | |||
| BC058333.1 | - | 1 | 790 | |||
| BC119602.2 | - | 1 | 765 | |||
| BC128564.1 | - | 1 | 760 | |||
| DQ201636.1 | - | 1 | 775 | |||

Nefm
Genomic Map