siRNA Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039181.1 | NP_001034270.1 | 3 | 962 | ||
| NM_001286395.1 | NP_001273324.1 | 4 | 578 | |||
| NM_008728.2 | NP_032754.2 | 3 | 962 | |||
| GenBank mRNA | AA124180.1 | - | 3 | 472 | ||
| AK079196.1 | - | 4 | 495 | |||
| AK166535.1 | - | 2 | 191 | |||
| BC055897.1 | - | 3 | 962 | 4019152 | ||
| BQ832991.1 | - | 2 | 131 | |||
| D78175.1 | - | 3 | 927 | |||
| U72676.1 | - | 2 | 165 | |||

Npr3
Genomic Map