siRNA Details
| Gene Symbol: Nxph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008752.2 | NP_032778.1 | 2 | 2221 | ||
| GenBank mRNA | AK038402.1 | - | 2 | 2222 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxph2| Gene Symbol: Nxph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008752.2 | NP_032778.1 | 2 | 2221 | ||
| GenBank mRNA | AK038402.1 | - | 2 | 2222 | ||