siRNA Details
| Gene Symbol: Oaz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301034.1 | NP_001287963.1 | 5 | 990 | ||
| NM_008753.4 | NP_032779.2 | 5 | 996 | |||
| GenBank mRNA | AK010615.1 | - | 5 | 967 | ||
| AK087942.1 | - | 2 | 2455 | |||
| AK146571.1 | - | 5 | 967 | |||
| AK147167.1 | - | 5 | 967 | |||
| AK151635.1 | - | 5 | 967 | |||
| AK159874.1 | - | 5 | 967 | |||
| AK160491.1 | - | 5 | 968 | |||
| AK160598.1 | - | 5 | 967 | |||
| AK169201.1 | - | 5 | 967 | |||
| AK181434.1 | - | 1 | 121 | |||
| BC002132.1 | - | 5 | 932 | |||
| BC024902.1 | - | 5 | 969 | |||
| BC046290.1 | - | 5 | 959 | |||
| BC060035.1 | - | 5 | 934 | |||
| BC091773.1 | - | 5 | 929 | |||
| BC094287.1 | - | 5 | 927 | 5257966 | ||
| BG296334.1 | - | 3 | 466 | |||
| U52822.1 | - | 5 | 898 | |||

Oaz1
Genomic Map