siRNA Details
| Gene Symbol: Odf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008757.3 | NP_032783.2 | 2 | 745 | ||
| GenBank mRNA | BC145758.1 | - | 2 | 745 | 40131061 | |
| EU791469.1 | - | 2 | 745 | |||
| X79446.1 | - | 2 | 749 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odf1| Gene Symbol: Odf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008757.3 | NP_032783.2 | 2 | 745 | ||
| GenBank mRNA | BC145758.1 | - | 2 | 745 | 40131061 | |
| EU791469.1 | - | 2 | 745 | |||
| X79446.1 | - | 2 | 749 | |||