siRNA Details
| Gene Symbol: Pabpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008774.3 | NP_032800.2 | 12 | 2096 | ||
| GenBank mRNA | AK044829.1 | - | 12 | 2093 | ||
| AK089747.1 | - | 12 | 2096 | |||
| AK133196.1 | - | 12 | 2097 | |||
| AK150839.1 | - | 12 | 2097 | |||
| AK152064.1 | - | 12 | 2064 | |||
| AK159703.1 | - | 12 | 2096 | |||
| AK159733.1 | - | 12 | 2096 | |||
| AK160883.1 | - | 12 | 2096 | |||
| AK160968.1 | - | 12 | 2095 | |||
| AK161123.1 | - | 12 | 2099 | |||
| AK161671.1 | - | 12 | 2095 | |||
| AK168466.1 | - | 12 | 2095 | |||
| BC003870.1 | - | 12 | 1715 | 3491915 | ||
| BC004587.1 | - | 11 | 1302 | |||
| BC011207.1 | - | 12 | 1853 | 4022169 | ||
| BC023145.1 | - | 12 | 1691 | 3988101 | ||
| BC046233.1 | - | 12 | 2086 | 6816124 | ||
| X65553.1 | - | 12 | 1882 | |||

Pabpc1
Genomic Map