siRNA Details
| Gene Symbol: Pcnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282992.1 | NP_001269921.1 | 6 | 962 | ||
| NM_008787.3 | NP_032813.3 | 6 | 962 | |||
| GenBank mRNA | AB207233.1 | - | 6 | 836 | ||
| BC085244.1 | - | 6 | 786 | |||
| BC100380.1 | - | 6 | 757 | |||
| BU518252.1 | - | 483 | ||||

Pcnt
Genomic Map