siRNA Details
| Gene Symbol: Pcp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008791.2 | NP_032817.1 | 3 | 397 | ||
| GenBank mRNA | BC061164.1 | - | 3 | 397 | 6593949 | |
| X17320.1 | - | 3 | 236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcp4| Gene Symbol: Pcp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008791.2 | NP_032817.1 | 3 | 397 | ||
| GenBank mRNA | BC061164.1 | - | 3 | 397 | 6593949 | |
| X17320.1 | - | 3 | 236 | |||